Learn More
Abcam Adenylate Cyclase Activity Assay
Product Code. 32115688 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115688 1 x 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115688 Supplier Abcam Supplier No. ab3236081x100Tests
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure Adenylate Cyclase activity in 1h 15m in Cell/Tissue Extracts with Adenylate Cyclase Activity Assay AB323608. Sensitivity <0.02 mU. Use with Microplate.
Measure Adenylate Cyclase activity in 1h 15m in Cell/Tissue Extracts with Adenylate Cyclase Activity Assay AB323608. Sensitivity <0.02 mU. Use with Microplate.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 100 Tests
Regulatory Status RUO