Learn More
Phenomenex™ Agela™ SPE Cartridge Frits
Product Code. 31744369 Shop All Phenomenex Products
Change view
Click to view available options
Volume (Metric):
12 mL
30 mL
Unit Size:
Pack of 100
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
31744369 30 mL Pack of 50
31743266 12 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31744369 Supplier Phenomenex™ Supplier No. AS030A
Expédition estimée: 19-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
PE Frit for SPE Tube
  • Brand: Agela
  • Technique: SP
  • Phase: No Phase
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Sorbent C18/SCX/SAX
Type Solid Phase Extraction Column
Volume (Metric) 30 mL