Learn More
Albaconazole, TRC
Product Code. 17937184
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17937184 5 mg 5mg
17968804 2.5 mg 2.5mg
17977834 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17937184 Supplier Toronto Research Chemicals Supplier No. TRCA5114505MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 187949-02-6
Molecular Formula C20 H16 Cl F2 N5 O2
Molecular Weight (g/mol) 431.82
IUPAC Name 7-chloro-3-[(2R,3R)-3-(2,4-difluorophenyl)-3-hydroxy-4-(1,2,4-triazol-1-yl)butan-2-yl]quinazolin-4-one
SMILES C[C@@H](N1C=Nc2cc(Cl)ccc2C1=O)[C@](O)(Cn3cncn3)c4ccc(F)cc4F
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C20H16ClF2N5O2/c1-12(28-11-25-18-6-13(21)2-4-15(18)19(28)29)20(30,8-27-10-24-9-26-27)16-5-3-14(22)7-17(16)23/h2-7,9-12,30H,8H2,1H3/t12-,20-/m1/s1
Formula Weight 431.0961
Chemical Name or Material Albaconazole