Learn More
Alpaca monoclonal [14A4] Anti-Mouse IgG2a/b antibody (ATTO 488), Abcam
Product Code. 32119156 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119156 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119156 Supplier Abcam Supplier No. ab325300100uL
 Restricted Item
Expédition estimée: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Alpaca Monoclonal Antibody
Recombinant secondary antibody suitable for ICC/IF.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Antigen Ighg (GCAA)
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone '14A4
Conjugate Atto 488
Formulation Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Ighg
Gene Accession No. P01863
Host Species Alpaca
Immunogen Ighg (GCAA)
Purification Method Size-exclusion chromatography
Quantity 100 μL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -80°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Ighg
Target Molecule Ighg (GCAA)