Learn More
alpha-Ketobutyric Acid, TRC
Product Code. 30136425
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136425 5 g 5g
30135473 100 g 100g
17953435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136425 Supplier Toronto Research Chemicals Supplier No. TRCK1753005G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 600-18-0
Molecular Formula C4 H6 O3
Molecular Weight (g/mol) 102.09
Synonym 2-Oxobutanoic Acid, 2-Oxobutyric Acid, 2-Ketobutanoic Acid, 2-Ketobutyric Acid, 2-Oxo-n-butyric Acid, 2-Oxobutanoic Acid, 2-Oxobutyric Acid, 3-Methylpyruvic Acid, NSC 60533, Propionylformic Acid, α-Ketobutyric Acid, α-Oxo-n-butyric Acid, α-Oxobutanoic Acid, α-Oxobutyric Acid, α-keto-n-Butyric Acid
IUPAC Name 2-oxobutanoic acid
SMILES CCC(=O)C(=O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C4H6O3/c1-2-3(5)4(6)7/h2H2,1H3,(H,6,7)
Formula Weight 102.0317
Chemical Name or Material alpha-Ketobutyric Acid