Learn More
Ansell AlphaTec™ 3000 Chemical Protective Suit Ultrasonically Welded and Taped, Model 122
Product Code. 18894518
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
18894518 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18894518 Supplier Ansell Supplier No. YE30T0012205
  • ONLINE EXCLUSIVE PRICE
    962.00€
    644.14€ / 20 pièces
    Économisez 317.86€
    33% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protective suit features a durable, comfortable, multi-layer chemical barrier and offers Type 3/4/5/6 protection.
  • Specialized protection: Made with durable multi-layer nonwoven laminate to help protect against various inorganic chemicals and biological hazards*, while ensuring lightweight comfort.
  • Enhanced features: Dual-zip systems and double cuffs enhance protective capabilities. Also features two-piece hood and an elasticated waist, sleeves and over-cuffs.
  • Improved visibility: High-visibility yellow exterior ensures greater wearer and workplace safety
  • Assured antistatic capabilities: Tested against EN 1149-5 antistatic standards.
  • *CE Category III Type 3/4/5/6 protection
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Hood Type Attached
Material Multi-layer Barrier Laminate Fabric
Size X-Large
Product Type Suit
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.