Learn More
Ansell AlphaTec™ 4000 Cape Hood
Product Code. 17170855
Change view
Click to view available options
Style: G40W507
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17170855 Style: G40W507
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17170855 Supplier Ansell Supplier No. GR40W0050700
Expédition estimée: 17-06-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hood
Size One Size
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Hook and Loop
Disposable Single-use
Seam Type Welded
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Style G40W507