Learn More
Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit
Product Code. 17609995 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
  • Three high positive control swabs
  • Three low positive control swabs
  • Three negative control swabs
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17609995
  • Three high positive control swabs
  • Three low positive control swabs
  • Three negative control swabs
1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17609995 Supplier Thermo Scientific™ Supplier No. RENCOV41001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
The Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test. When used for quality control in the test, they are useful as an aid in verifying test and operator performance.
This product has been discontinued and is no longer available for purchase.
The Thermo Scientific™ Renvo™ SARS-CoV-2 Control Kit provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test. When used for quality control in the test, they are useful as an aid in verifying test and operator performance. Intended use:
For environmental surveillance use only and not for diagnostic purposes.
Contents:
  • SARS-CoV-2 high positive control swab (3): DNA-based synthetic oligo dried onto a swab above the limit of detection of the test.
  • SARS-CoV-2 low positive control swab (3):DNA-based synthetic oligo dried onto a swab above the limit of detection of the test.
  • SARS-CoV-2 negative control swab (3): Buffer solution dried onto a swab.
We recommend running a SARS-CoV-2 positive and negative control:
  • Once for each new lot or shipment of kits received
  • Once for each new operator
TRUSTED_SUSTAINABILITY
Spécification
Description  Provides positive and negative control swabs for use with the Thermo Scientific™ Renvo™ SARS-CoV-2 Test
Includes
  • Three high positive control swabs
  • Three low positive control swabs
  • Three negative control swabs