Learn More
Ansell AlphaTec™ Airline Passthrough without Connector
Product Code. 18813796
Change view
Click to view available options
Includes:
Without Connector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes
18813796 Without Connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18813796 Supplier Ansell Supplier No. AC01P0002200N00
  • ONLINE EXCLUSIVE PRICE
    17239.10€
    15121.98€ / 18 pièces
    Économisez 2117.12€
    12% de réduction
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Provides external air supply to AlphaTec reusable gas-tight suits, for breathing air as well as suit ventilation.
  • For use with external air supply: Useful during suit decontamination
  • Large thumbwheel: Good grip, easy adjustments
  • Compatible with all SCBA brands: Improved practicality and convenience
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 100 L/min.
Material Glass Fiber Reinforced PPS, Polyamide, Anti-static Polyester
Includes Without Connector
Product Type Air Supply Connector