Learn More
Brady™ Aluminum: ISO Safety Sign - Wear safety footwear
Product Code. 15684477 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15684477 200 mm Each
15674477 100 mm Each
15694477 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15684477 Supplier Brady™ Supplier No. 819025
  • ONLINE EXCLUSIVE PRICE
    16.00€
    13.51€ / 1 pièces
    Économisez 2.49€
    15% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Sign compliant to ISO 7010.
  • Aluminum
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M008-Dia 200-AL-CRD/1
Material Aluminum
Quantity 1 Ea.
Diameter (Metric) 200 mm
For Use With (Application) Indoor and Outdoor
Thickness 1.016 mm
Temperature Range (Metric) -40°C to 140°C
Shape Round
Languages Global
Color White on Blue