Learn More
Brady™ Aluminum Warning Sign: Explosive Atmosphere
Product Code. 11730943 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
315 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
11730943 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11730943 Supplier Brady™ Supplier No. 810890
  • ONLINE EXCLUSIVE PRICE
    32.90€
    27.81€ / 1 pièces
    Économisez 5.09€
    15% de réduction
Expédition estimée: 14-12-2026