Learn More
WTW™ Ammonia Gas-sensitive Combination Electrode
Product Code. 10461402
Change view
Click to view available options
Quantity:
3 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10461402 3 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10461402 Supplier WTW™ Supplier No. 106395
  • ONLINE EXCLUSIVE PRICE
    1330.00€
    1210.87€ / 1 pièces
    Économisez 119.13€
    8% de réduction
Expédition estimée: 02-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The half cells of the 500 series for which you will need a reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated reference electrode).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Membrane Heads
Quantity 3 Pk.