Learn More
Interscience™ Anabac Deodorizing Capsules
Product Code. 10422084
Change view
Click to view available options
Scent:
Apple
Citrus
Fresh Scent
Color:
Clear
Green, White
Yellow
Appearance:
Clear Amber
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent Color Appearance Sterility Unit Size
10422084 Fresh Scent Clear Yellow Sterile Each
15388927 Apple Green, White Clear Amber Sterile Pack of 100
16825923 Citrus Yellow Yellow Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10422084 Supplier Interscience™ Supplier No. 320100
  • ONLINE EXCLUSIVE PRICE
    123.00€
    99.28€ / 1 pièces
    Économisez 23.72€
    19% de réduction
 Restricted Item
Expédition estimée: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Placed in an autoclave, Anabac capsules provide a discreet way to perfume the laboratory.
  • Capsule dissolves during an autoclaving destruction cycle
  • Capsule melts completely without leaving residue
  • Eliminates unpleasant odors that can emerge from the autoclave
Specifications
Product Type Autoclave Capsule
Color-coded Yes
Form Capsules
Handle Material Plastic
Handle Type Cap
Scent Fresh Scent
Sterility Sterile
Appearance Yellow
Autoclavable Autoclavable
Color Clear
Disposable Single-use
Filtered Non-filtered
For Use With (Application) Refreshes and Perfumes Air Where Autoclave Is Located
For Use With (Equipment) Autoclave
Quantity 100 Capsules/Pk.
Show More Show Less
compliance-icons
Product Identifier
  • Anabac deodorant
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.