Learn More
Gibco™ Animal-Free Murine Dendritic Cell Cytokine Package, PeproTech®
Product Code. 17846857 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17846857 1 pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17846857 Supplier Gibco™ Supplier No. AFMDC
  • ONLINE EXCLUSIVE PRICE
    1000.00€
    940.00€ / 1 pièces
    Économisez 60.00€
    6% de réduction
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Animal-Free Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
The Animal-Free Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
The package contains the following two vials:
• 100 μg Animal-Free Murine IL-4 (Cat. No. AF-214-14-100UG)
• 100 μg Animal-Free Murine GM-CSF (Cat. No. AF-315-03-100UG)
IL-4 is a pleiotropic cytokine that regulates diverse T- and B-cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Functional Assay
Formulation Lyophilized
Product Type Dendritic Cell Cytokine Package
Quantity 1 pack
Species Murine