Learn More
SI Analytics™ Anodized Aluminum Measuring Stand
Product Code. 10253271 Shop All SI Analytics Products
Change view
Click to view available options
Material:
Anodized Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10253271 Anodized Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10253271 Supplier SI Analytics™ Supplier No. 285410502
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
SI Analytics™ Anodized Aluminum Measuring Stand is for use with capillary viscometers and in thermostatic baths.
  • Measuring stand anodized aluminum AVS™/S
  • Material: anodized aluminum
  • Product Type: measuring stand
TRUSTED_SUSTAINABILITY
Specifications
Product Type Measuring Stand
For Use With (Equipment) Capillary Viscometer
Material Anodized Aluminum
For Use With (Application) Thermostatic Baths from -80 to 80°C
Temperature Range (Metric) -80°C to +80°C