missing translation for 'onlineSavingsMsg'
Learn More

IFN-γ Rat anti-Mouse, APC, Clone: XMG1.2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554413

Nacres classification code: NA.46

 View more versions of this product

Product Code. 15867678

  • ONLINE EXCLUSIVE PRICE
    381.00€
    380.92€ / 1 pièce
    Économisez 0.08€
    0.02% de réduction

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The XMG1.2 antibody reacts with mouse interferon-γ (IFN-γ) protein. IFN-γ is a pleiotropic cytokine, of approximately 15-17kDa, involved in the regulation of the immune response. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN-γ production is associated with the Th-1 differentiation. This is a neutralizing antibody

Host Species: Rat
Clone: XMG1.2
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse IFN-γ

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFN-γ
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse IFN-γ
0.1 mg
Immunology
Mouse
IgG1 κ
Flow Cytometry
XMG1.2
APC
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.