Learn More
Apitolisib, MedChemExpress
Product Code. 16496126
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496126 100 mg 100mg
16445588 10 mg 10mg
16418287 5 mg 5mg
16411757 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496126 Supplier Medchem Express Supplier No. HY13246
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Apitolisib (GDC-0980; GNE 390; RG 7422) is a selective, potent, orally bioavailable Class I PI3 kinase and mTOR kinase (TORC1/2) inhibitor with IC50s of 5 nM/27 nM/7 nM/14 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ, and with a Ki of 17 nM for mTOR.
Apitolisib (GDC-0980
Specifications
Chemical Name or Material Apitolisib
Physical Form Solid
CAS 1032754-93-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H30N8O3S
For Use With (Application) Cancer-Kinase/protease
Synonym GDC-0980 GNE 390 RG 7422
Solubility Information DMSO : 14.29 mg/mL (28.66 mM; Need ultrasonic)
SMILES C[C@@H](C(N1CCN(CC1)CC2=C(C3=NC(C4=CN=C(N=C4)N)=NC(N5CCOCC5)=C3S2)C)=O)O
Molecular Weight (g/mol) 498.6
Formula Weight 498.6
Percent Purity 97.88%
Grade Research