Learn More
Thermo Scientific™ Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Product Code. 17946982
Change view
Click to view available options
Type:
Electrode Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17946982 Electrode Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17946982 Supplier Thermo Scientific™ Supplier No. 2111XPEK
  • ONLINE EXCLUSIVE PRICE
    1470.00€
    1197.73€ / 1 kits
    Économisez 272.27€
    18% de réduction
 Restricted Item
Expédition estimée: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Adapt your sodium analyzer to specific needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzer.
Adapt your sodium analyzer to your specific analytical needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer.
Warranty: Out of box
TRUSTED_SUSTAINABILITY
Specifications
Description 2111LL Electrode Kit
Type Electrode Kit
For Use With (Equipment) Orion™ 2111XP Sodium Analyzer
Includes 2111LL Electrode Kit - includes sodium electrode, reference electrode and 2 oz. reference electrode filling solution
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.