Learn More
Sartorius Arium™ Advance L Filter Housing
Product Code. 16877286 Shop All Sartorius Products
Change view
Click to view available options
Description:
Double
Single
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
16877286 Single Each
16887286 Double Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16877286 Supplier Sartorius Supplier No. H2OAFHL1
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Arium™ Advance L Filter Housing for flexible configurations of filters.
  • The Arium™ Advance L filter housing as either one-stage or two-stage allows for flexible configurations of filters for pre-treatment of water according to your system requirements.
TRUSTED_SUSTAINABILITY
Specifications
Description Single
Feedwater Source Potable Tap Water
Product Water Type Reverse Osmosis (Type 3)
Type Filter Housings and Holders
Material Polypropylene Housing