Learn More
Asivatrep, MedChemExpress
Product Code. 16485338
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485338 10 mg 10mg
16458057 5 mg 5mg
16451537 50 mg 50mg
16445976 100 mg 100mg
16441299 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485338 Supplier Medchem Express Supplier No. HY12777
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Asivatrep (PAC-14028) is a potent and selective transient receptor potential vanilloid type I (TRPV1) antagonist.
Asivatrep (PAC-14028) is a potent and selective transient receptor potential vanilloid type I (TRPV1) antagonist.
Specifications
Chemical Name or Material Asivatrep
CAS 1005168-10-4
Color White
Purity Grade Notes Research
Molecular Formula C21H22F5N3O3S
Quantity 10 mg
Synonym PAC-14028
Solubility Information DMSO : 50 mg/mL (101.74 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C1=CC(F)=C(NS(=O)(C)=O)C(F)=C1)C)/C=C/C2=CC=C(C(F)(F)F)N=C2CCC
Molecular Weight (g/mol) 491.47
Formula Weight 491.47
Percent Purity 95.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation