Learn More
Saint-Gobain AstiPure™ MF1 Series Flare Tool
Click to view available options
For Use With:
For 10 x 13mm dia. tubing
For 8 x 10mm dia. tubing
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | ISO 9001 |
| For Use With | For 10 x 13mm dia. tubing |
| Packaging Quantity | 1 Each |
| Includes | Nut |
| Product Type | Flare Tool |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction