Learn More
Saint-Gobain AstiPure™ V3V Series PFA 3-Way Stopcock Valve
Click to view available options
Diameter (Metric) Inner:
6 mm
8 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 6 mm |
| Material | PFA |
| Packaging Quantity | 1 Each |
| Product Type | 3-Way Stopcock Valve |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction