Learn More
Specac™ Atlas™ Hydraulic Press Automatic 15T
Product Code. 15270348
Encompass_Preferred
Change view
Click to view available options
Type:
Atlas Hydraulic Press Automatic 15T
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15270348 Atlas Hydraulic Press Automatic 15T Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15270348 Supplier Specac™ Supplier No. GS25430
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
This Atlas Power 15T Press is a power assisted hydraulic press that operates at up to 15 Tons. It has been designed to handle a wide variety of pressing applications including XRF and IR sample preparation. This press has a microprocessor controlled pressure application and release, Touch Screen operation and can accommodate samples up to 200mm in diameter. They can also be used with Specac Heated Platens GS15515 and Specac Evacuable Pellet Dies. Supplied with UK/EU Plug.
Specifications
Type Atlas Hydraulic Press Automatic 15T