Learn More
Invitrogen™ Attune™ NxT and CytPix Waste Bottle
Product Code. 17148375
Change view
Click to view available options
Quantity:
1 bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17148375 1 bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17148375 Supplier Invitrogen™ Supplier No. 100085928CST
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
This is a 1.9-L replacement bottle for the waste tank on the Attune NxT Acoustic Focusing Cytometer.
  • This is a 1.9-L replacement bottle for the waste tank on the Attune NxT Acoustic Focusing Cytometer.
  • The bottle is designed for easy user replacement and with more robust fittings to prevent breakage.
  • This bottle replaces discontinued Cat. No. 100022156.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 Bottle
For Use With (Equipment) Attune NxT
Includes Waste Bottle
Product Line FPL
Quantity 1 bottle
Shipping Condition Room temperature
Product Type Flow Cytometer Accessory
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.