Description
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
Match Your CETAC Sample Tube Rack to Your Laboratory’s Needs
- 21-position sample tube rack
- 24-position sample tube rack
- 40-position sample tube rack
- 60-position sample tube rack
- 90-position sample tube rack
- Extended Rack for CETAC ASX-520 Autosampler (for use with Thermo Scientific™ iCAP™ Q Series ICP-MS systems)
Specifications
Specifications
| For Use With (Equipment) | iSC-65,CETAC, ESI |
| Type | Autosampler Sample Tube Racks |
| Includes | 60-Position Sample Tube Rack |
Product Title
¿Detecta una oportunidad de mejora?