Learn More
Zeiss™ Axioscope 5 Upright Microscope
Product Code. 17362810
Change view
Click to view available options
For Use With (Application):
Transmitted Light Brightfield and DIC
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17362810 Transmitted Light Brightfield and DIC 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17362810 Supplier Zeiss™ Supplier No. 4900400048000
  • ONLINE EXCLUSIVE PRICE
    45190.00€
    30494.00€ / 1 kits
    Économisez 14696.00€
    32% de réduction
View all promotions
 Restricted Item
Expédition estimée: 26-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Flexible upright microscope ideal for biomedical routine & research. Configurable for the clinical environment. System is configured for transmitted brightfield & DIC microscopy with phototube. Use in combination with Axiocam 208 color for a smart standalone microscope system.
  • The Axioscope 5 is a flexible manual upright microscope that can be used for a for a range of transmitted light observations and also a fluorescence model is available. The possible contrast methods are brightfield, darkfield, phase contrast, PlasDIC, DIC, fluorescence,
    and transmitted/reflected light polarization depending on the configuration. This allows the microscope to be used for a range of applications in biology research, human and veterinary medicine, microbiology, plant science & botany and forensics.
  • The microscope body is powder coated durable metal, heat treated and resistant to standard reagents used for staining, organic solvents used for cleaning of lenses and disinfectants.
  • All optical parts including objectives, eyepieces, lenses, prisms, have anti-reflective and anti-fungal coating.
  • The microscope offers a coaxial z-drive with coarse and fine focus. Fine focus knob left and right.The focus range is 24 mm. The microscope includes an adjustable focus stop. The coarse focus provides movements of 4 mm/rotation, the fine focus comes with 400μm/rotation and with a graduation in micrometers (4μm).
  • The microscope comes with a manual revolving nosepiece with 6 encoded positions, suitable for reading objective position for light intensity manager functionality and for scaling information.
  • The microscope provides an energy saving and illumination life time extending Eco-mode function. When active, your microscope automatically goes to standby after being idle for 15 minutes, with a click of any button the microscope system is instantly on again.
  • The microscope body is equipped with two integrated, ergonomically positioned “snap image buttons” close to the focus drive on both sides of the microscope body to minimize hand movements and to optimally support left and right handed users. The snap image button allows to acquire images and videos directly from the stand (with Axiocam 208/202 in stand-alone mode and with Labscope; with any Axiocam with ZEN).
Specifications
For Use With (Application) Transmitted Light Brightfield and DIC