Learn More
Azaperone 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30326360
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326360 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326360 Supplier Dr. Ehrenstorfer Supplier No. DREA10340510AL1000
 Restricted Item
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Azaperone
CAS 1649-18-9
Analyte or Component Names Azaperone
Shipping Condition +20°C
Molecular Formula C19 H22 F N3 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Azaperone, 1-(4-Fluorophenyl)-4-[4-(pyridin-2-yl)piperazin-1-yl]butan-1-one, 1-Butanone, 1-(4-fluorophenyl)-4-[4-(2-pyridinyl)-1-piperazinyl]-, Butyrophenone, 4'-fluoro-4-[4-(2-pyridyl)-1-piperazinyl]- (6CI,8CI), 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, Azaperone, Fluoperidol, NSC 170976, R-1929, Stresnil, Suicalm
InChI Formula InChI=1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2
SMILES Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3
IUPAC Name 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 327.4
Quantity 1 mL
Formula Weight 327.1747 g/mol