Learn More
Azelastine N-Oxide (Mixture of Diastereomers), TRC
Product Code. 30041071
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041071 5 mg 5mg
30040432 50 mg 50mg
30041387 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041071 Supplier Toronto Research Chemicals Supplier No. TRCA8082605MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 640279-88-5
Molecular Formula C22 H24 Cl N3 O2
Molecular Weight (g/mol) 397.9
Synonym 1(2H)-Phthalazinone, 4-[(4-chlorophenyl)methyl]-2-(hexahydro-1-methyl-1-oxido-1H-azepin-4-yl)-, Azelastine N-Oxide
IUPAC Name 4-[(4-chlorophenyl)methyl]-2-(1-methyl-1-oxidoazepan-1-ium-4-yl)phthalazin-1-one
SMILES C[N+]1([O-])CCCC(CC1)N2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C22H24ClN3O2/c1-26(28)13-4-5-18(12-14-26)25-22(27)20-7-3-2-6-19(20)21(24-25)15-16-8-10-17(23)11-9-16/h2-3,6-11,18H,4-5,12-15H2,1H3
Formula Weight 397.1557
Percent Purity >95%
Chemical Name or Material Azelastine N-Oxide (Mixture of Diastereomers)
Grade HPLC