Learn More
J.T. Baker™ BAKER spe-12G Column Processor, Glass Design
Product Code. 15597644 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15597644 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597644 Supplier J.T. Baker™ Supplier No. 699800
  • ONLINE EXCLUSIVE PRICE
    3565.69€
    2451.06€ / 1 pièces
    Économisez 1114.63€
    31% de réduction
Expédition estimée: 19-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Offers the flexibility of processing up to 12 SPE devices of different heights, diameters, or formats at the same time. J.T. Baker™ BAKER spe-12G Column Processor, Glass Design is a complete 12-port system that comes complete with a glass vacuum basin and sample collection rack.
The design of the BAKER spe-12G Column Processor, Glass Design is familiar throughout the industry, and supports all devices and accessories with luer-type fittings (e.g. BAKERBOND spe, BAKERBOND Speedisk™ columns, and BAKERBOND Speedisk™ extraction disks).
The J.T. Baker™ BAKER spe-12G is a 12-port system, suitable for processing up to 12 SPE columns at the same time. The processor comes complete with:
  • glass vacuum basin
  • cover with luer fittings and gasket
  • individual flow control stopcocks
  • stainless steel needles
  • glass sample collection rack
  • height-adjustable shelves
  • vacuum gauge/controller
Specifications
For Use With (Application) Solid Phase Extraction
For Use With (Equipment) Extraction Columns and Discs with Luer-Type Fittings, Such as BAKERBOND Spe, BAKERBOND Speedisk Colu
Type SPE Vacuum Processor
Material Glass vacuum chamber and sample collection rack, polyamide lid, PTFE luer connectors/stopcocks, polyethylene/neoprene gaskets
Quantity 1 Each