Learn More
Avantor™ BAKERBOND Speedisk™ Extraction Disks
Product Code. 15568904 Shop All J.T.Baker Products
Change view
Click to view available options
Type:
Octadecyl (C18 XF) Extraction Disk
Octyl (C8) Extraction Disk
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15568904 Octyl (C8) Extraction Disk Each
10406423 Octadecyl (C18 XF) Extraction Disk Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15568904 Supplier Avantor™ Supplier No. 805706
  • ONLINE EXCLUSIVE PRICE
    614.51€
    465.05€ / 1 pièces
    Économisez 149.46€
    24% de réduction
 Restricted Item
Expédition estimée: 25-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) Diquat, Paraquat, EPA Method 549.1
For Use With (Equipment) Expanded Extraction Station (8095-06)
Sorbent C18 XF
Type Octyl (C8) Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diisopropyl ether