Designed for laboratories, classrooms and light duty applications where safe ventilation is required. Constructed of powder-coated steel, Basic Hoods are economical and have a smaller overall height and depth than traditional hoods so they fit in areas with low ceilings or narrow aisles.
For areas with low ceilings or narrow aisles, choose the Basic™ Laboratory Hood when outfitting your lab.
By-pass airflow design
Durable epoxy-coated 16 gauge steel exterior and interior resists chemical exposure
LED lighting
Vertical-rising tempered safety glass sash
Adjustable two-piece baffle
Curved air foil
Right side fixture panel factory-prepared to accept up to three service fixtures, one electrical duplex and one airflow monitor (kits sold separately)
Right interior wall factory-prepared for one cupsink and turret (cupsink and turret kit sold separately)
Epoxy-coated steel 10.31 in. OD exhaust connection
Order Info
Exterior D x H: 25 x 48 in. (64 x 122 cm). Add 5 in. (12.7 cm) to height for models with built-in blower. Available with or without blower. Blower may be mounted in hood liner, above hood, or at remote roof location.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy.