missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%

Product Code. 11410857
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410857 5 g 5g
11344599 250 mg 250mg
11400857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410857 Supplier Thermo Scientific Alfa Aesar Supplier No. B23244.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol

Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52746-49-3
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.48
MDL Number MFCD00149310
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
PubChem CID 54669730
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in water.
IUPAC Name disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
Formula Weight 536.48 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.