Learn More
Dräger™ Battery/Charging Accessories - Multinational Power Supply
Product Code. 15666544
Change view
Click to view available options
Type:
Power Supply
Single Charger
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15666544 Single Charger Each
10312859 Power Supply Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15666544 Supplier Dräger™ Supplier No. 8316994
  • ONLINE EXCLUSIVE PRICE
    246.02€
    192.44€ / 1 pièces
    Économisez 53.58€
    21% de réduction
Expédition estimée: 11-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W x H) 6 x 6 x 6 in. (15.24 x 15.24 x 15.24cm)
Type Single Charger
Height (English) 6 in.
Height (Metric) 15.24 cm
Length (English) 6 in.
Length (Metric) 15.24 cm
Voltage 100/240VAC
Width (English) 6 in.
Width (Metric) 15.24 cm