Learn More
Brady™ BBP12 Label Printer Kit EU with Cutter and Unwinder
Product Code. 15717042 Shop All Brady Products
Change view
Click to view available options
BBP12 Label Printer Kit EU; Print Speed: 101.6 mm/s; Resolution: 300 dpi; Print Technology: Thermal Transfer; Weight (Metric): 2.3 kg; Color Capability: Monochrome
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15717042 BBP12 Label Printer Kit EU; Print Speed: 101.6 mm/s; Resolution: 300 dpi; Print Technology: Thermal Transfer; Weight (Metric): 2.3 kg; Color Capability: Monochrome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15717042 Supplier Brady™ Supplier No. 195966
  • ONLINE EXCLUSIVE PRICE
    929.00€
    723.45€ / 1 pièces
    Économisez 205.55€
    22% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
The BBP12 Label Printer is a compact, reliable, and affordable entry level thermal transfer benchtop printer ideal for low volume printing on lots of different materials.
The BBP™12 gives you the versatility to print all your basic label types from one machine, saving costs, reducing downtime and simplifying your labeling.
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts for general identification, wire marking, asset tracking and more
  • Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Comes with cutter, unwinder, and free Brady™ Workstation Basic Design Software Suite
Specifications
Display Type Color LCD
Print Technology Thermal Transfer
Width (English) 8 in.
Width (Metric) 203 mm
Height (English) 7.5 in.
Height (Metric) 191 mm
Depth (English) 10 in.
Depth (Metric) 259 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes BBP12 Printer with Pre-installed Cutter, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Brady™ Basic Suite
Languages Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
Memory 128 MB (Flash), 64 MB (SDRAM), SD Flash Memory Card Up To 32 GB
Model BBP12
Print Speed 101.6 mm/s
Resolution 300 dpi
Connector Types Ethernet, Serial, USB
Operating System Windows™ 7, Windows™ 8, Windows™ 10
Temperature Range (Metric) 5°C to 40°C
Weight (English) 5.1 lb.
Weight (Metric) 2.3 kg
For Use With (Application) Barcode Labeling, Laboratory Labeling, Wire and Cable Labeling
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number BBP12-EU-U-CUTTER