Learn More
BD CBA Human Inflammatory Cytokine Kit
Product Code. 15806708 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15806708 80 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15806708 Supplier BD Supplier No. 551811
 Restricted Item
Expédition estimée: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Includes
Reagents for capture and detection of IL-8, IL-1β, IL-6, IL-10, TNF, and IL-12p70, wash buffer, assay diluent, serum enhancement buffer, cytometer setup reagents
Optimized for analysis of specific proteins in tissue culture supernatants, EDTA treated plasma, and serum samples
  • Can be used to quantitatively measure interleukin-8 (IL-8), interleukin-1β (IL-1β), interleukin-6 (IL-6), interleukin-10 (IL-10), tumor necrosis factor (TNF), and interleukin-12p70 (IL-12p70) protein levels in a single sample
  • Detection reagent provided in the kit is a mixture of phycoerythrin (PE)–conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
  • Provides sufficient reagents for analysis of 80 samples
For research use only. Not for use in diagnostic or therapeutic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-8, IL-1beta, IL-6, IL-10, TNF and IL-12p70
For Use With (Application) For analysis of specific chemokines in Tissue Culture supernatants, EDTA-treated Plasma (EDTA-plasma) and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
Reactivity Human