Learn More
BD Cell Viability Solution
Product Code. 15878458 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878458 100 Tests Each
15868458 500 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15878458 Supplier BD Supplier No. 555816
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ready-to-use solution of the nucleic acid dye, 7-Amino-actinomycin D (7-AAD) that can be used in place of propidium iodide (PI) for the exclusion of nonviable cells in flow cytometric assays
  • Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
  • Does not require dilution
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Quantity 100 Tests