Learn More
Bel-Art™ Poxygrid™ Imhoff Cone Racks
Product Code. 13432199
Change view
Click to view available options
Length (Metric):
44.45 cm
No. of Places:
3
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) No. of Places Unit Size
13432199 44.45 cm 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13432199 Supplier Bel-Art™ Supplier No. F389930003
  • ONLINE EXCLUSIVE PRICE
    152.00€
    100.41€ / 1 pièces
    Économisez 51.59€
    33% de réduction
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Lightweight Poxygrid racks support three or four Imhoff settling cones in a single row for clearer viewing
  • Cone tips are held up off the benchtop for easier viewing; allows plug removal if required
  • Wire frames are epoxy coated for durability and easy cleaning
  • Wide base provides stability without crowding the benchtop
Specifications
Volume (Metric) 3 Places
No. of Places 3
Tip Style Cone
Type Imhoff Settling Cone Rack
Dimensions (L x W x H) 17.5 x 6.75 x 16 in. (44.45 x 17.14 x 40.6 cm)
Length (English) 17.5 in.
Length (Metric) 44.45 cm
Width (English) 6.75 in.
Width (Metric) 17.14 cm
Height (English) 16 in.
Height (Metric) 40.6 cm