Learn More
Bel-Art™ Slide Tray Cabinet
Product Code. 10530754
Change view
Click to view available options
Depth (English):
14 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (English) Unit Size
10530754 14 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10530754 Supplier Bel-Art™ Supplier No. H445830000
  • ONLINE EXCLUSIVE PRICE
    356.00€
    339.03€ / 1 pièces
    Économisez 16.97€
    4% de réduction
Expédition estimée: 01-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Compact cabinet to organize and store microscope slides
  • Small footprint cabinet can be placed on the benchtop, making slides easily accessible
  • Dust-free storage
  • Holds 240 slides in 12 microscope trays (Mfr. No. H44581-0000, not included)
  • Polypropylene outer shell; polystyrene tray rails
  • Transparent vertically sliding acrylic door with door knob
  • 35.6L x 20.8W x 25.4H cm (14 x 83/16 x 10 in.)
Specifications
Type Slide Storage Cabinet
Slide Capacity 240
Depth (English) 14 in.
Depth (Metric) 356 mm
Device Holds 240
For Use With (Equipment) Microscope
Height (English) 10 in.
Height (Metric) 254 mm
Material Polypropylene, Polystyrene
Width (English) 5.18 in.
Width (Metric) 131 mm