Learn More
Benzoic anhydride, 95%, BLD Pharm™
Product Code. 31331552
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31331552 25 g
31330502 500 g
31328442 100 g
31326132 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31331552 Supplier BLD Pharm Supplier No. BD11583325g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 93-97-0
Molecular Formula C14H10O3
MDL Number MFCD00003073
Quantity 25 g
SMILES O=C(OC(C1=CC=CC=C1)=O)C2=CC=CC=C2
Molecular Weight (g/mol) 226.23
Percent Purity 95%
Chemical Name or Material Benzoic anhydride