Learn More
Benztropine mesylate, MedChemExpress
Product Code. 16472209
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472209 Supplier Medchem Express Supplier No. HYB0520A
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Benztropine mesylate (Benzatropine mesylate) is an orally active centrally acting anticholinergic agent that can be used for Parkinson's disease research. Benztropine mesylate is an anti-histamine agent and a dopamine re-uptake inhibitor. Benztropine mesylate is also a human D2 dopamine receptor allosteric antagonist. Benztropine mesylate also has anti-CSCs (cancer stem cells) effects.
Benztropine mesylate (Benzatropine mesylate) is an orally active centrally acting anticholinergic agent that can be used for Parkinson's disease research. Benztropine mesylate is an anti-histamine agent and a dopamine re-uptake inhibitor. Benztropine mesylate is also a human D2 dopamine receptor allosteric antagonist. Benztropine mesylate also has anti-CSCs (cancer stem cells) effects.
Specifications
Chemical Name or Material Benztropine mesylate
CAS 132-17-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H29NO4S
Quantity 1 g
Synonym Benzatropine mesylate Benzotropine mesylate Benztropine methanesulfonate
Solubility Information H2O : 100 mg/mL (247.81 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (247.81 mM; Need ultrasonic)
SMILES CN1[C@H]2CC(OC(C3=CC=CC=C3)C4=CC=CC=C4)C[C@@H]1CC2.CS(=O)(O)=O
Molecular Weight (g/mol) 403.54
Formula Weight 403.54
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration