missing translation for 'onlineSavingsMsg'
Learn More

Benzyl propargyl ether, 97%

Product Code. 15430348
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430348 5 g 5g
15440348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430348 Supplier Thermo Scientific Alfa Aesar Supplier No. H53481.06
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy

CAS: 4039-82-1 | C10H10O | 146.189 g/mol

Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

Solubility
Soluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4039-82-1
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00483987
InChI Key PAQVEXAFKDWGOT-UHFFFAOYSA-N
Synonym benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
PubChem CID 6917484
IUPAC Name prop-2-ynoxymethylbenzene
SMILES C#CCOCC1=CC=CC=C1

Specifications

Density 1.011
Boiling Point 101°C to 101°C (18 mmHg)
Linear Formula C6H4CH2OCH2C≡CH
Refractive Index 1.52
Quantity 5 g
Solubility Information Soluble in chloroform.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material Benzyl propargyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.