Learn More
Benzyltriethylammonium bromide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 5197-95-5 |
| Molecular Formula | C13H22BrN |
| MDL Number | MFCD00011822 |
| Quantity | 500 g |
| SMILES | CC[N+](CC)(CC1=CC=CC=C1)CC.[Br-] |
| Molecular Weight (g/mol) | 272.22 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzyltriethylammonium bromide |