Learn More
beta Max-ES liquid scintillation cocktail, MP Biomedicals™
Product Code. 15782242 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 gal.
Unit Size:
3.79 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15782242 1 gal. 3.79 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782242 Supplier MP Biomedicals Supplier No. 0188002001
  • ONLINE EXCLUSIVE PRICE
    344.00€
    327.60€ / 3.79L
    Économisez 16.40€
    4% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Chemical Name or Material Beta Max-ES liquid scintillation cocktail
Density 0.9 to 1.0g/mL at 20°C
Quantity 1 gal.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Betamax ES
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302+H332-Harmful if swallowed or if inhaled
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .