Learn More
CleanSpace Technology™ BioHood
Product Code. 17465382 Shop All CleanSpace Technology Products
Change view
Click to view available options
BioHood; Material: Flash Spun Polyethylene; Facepiece Type: Polycarbonate Visor; Color: White; For Use With (Application): Protection From Splash and Airborne Contaminents; Sterility: Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17465382 BioHood; Material: Flash Spun Polyethylene; Facepiece Type: Polycarbonate Visor; Color: White; For Use With (Application): Protection From Splash and Airborne Contaminents; Sterility: Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17465382 Supplier CleanSpace Technology™ Supplier No. CS3024
  • ONLINE EXCLUSIVE PRICE
    129.93€
    100.00€ / 1 pièces
    Économisez 29.93€
    23% de réduction
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The BioHood System is for use with the HALO and is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms.
The BioHood System is compatible with the CleanSpace™ HALO Respirator. It is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms, providing eye and head protection.
  • DuPont™ Tyvek™ material and seam technology
  • Light and durable
  • Blood and fluid resistant
  • Only PAPR hood design to completely cover the respirator when worn, minimizing device contamination
  • Can be easily removed while still wearing the respirator to protect healthcare professionals when doffing
  • Broad field of vision and unique design that secures the visor to eliminate slippage and movement
  • No belts, hoses, fogging or heat
  • Comfortable air flow with no breathing resistance
  • NIOSH and CE Mark approved
  • BioHood Parts: The BioHood (CS3024), Bio HEPA P3/T3 (CS3025), Bio Mask Exhalation Adaptor (CS3026) - reusable and attaches to CleanSpace™ HALO Face Mask (face mask sold separately)
Specifications
Color White
Material Flash Spun Polyethylene
Facepiece Type Polycarbonate Visor
For Use With (Application) Protection From Splash and Airborne Contaminents
For Use With (Equipment) CS3025, CS3027
Latex Free Yes
Sterility Non-sterile
Dimensions (L x W x H) 31 x 14 x 10 cm