Learn More
MedChemExpress Biotin LC hydrazide
Product Code. 16456357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456357 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16456357 Supplier MedChemExpress Supplier No. HY101885
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Biotin LC hydrazide is a long chain protein modification reagent, which can transform periodate-oxidized glycoproteins.
Specifications
Quantity 5 mg
Product Type Biochemical Assay Reagents
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 109276-34-8
Molecular Weight (g/mol) 371.5 g/mol
Solubility DMSO : 31 mg/mL (83.45 mM; Need ultrasonic)
Synonym Biotinamidocaproyl hydrazide ∣ BACH ∣ (+)-Biotinamidohexanoic Acid hydrazide ∣ Biotin Aminocaproyl Hydrazide