Learn More
(+)-Biotin-ONP, TRC
Specifications
Specifications
| Chemical Name or Material | D-Biotin 4-Nitrophenyl Ester |
| CAS | 33755-53-2 |
| Molecular Formula | C16H19N3O5S |
| Quantity | 25 mg |
| Synonym | (+)-Biotin p-nitrophenyl ester, (+)-Biotin 4-nitrophenyl ester, [3aS-(3aalpha,4beta,6aalpha)]-hexahydro-2-oxo-1H-Thieno[3,4-d]imidazole-4-pentanoic acid nitrophenyl ester, d-Biotin p-nitrophenyl ester, Biotin-Onp |
| InChI Formula | InChI=1S/C16H19N3O5S/c20-14(24-11-7-5-10(6-8-11)19(22)23)4-2-1-3-13-15-12(9-25-13)17-16(21)18-15/h5-8,12-13,15H,1-4,9H2,(H2,17,18,21)/t12-,13-,15-/m0/s1 |
| SMILES | [O-][N+](=O)c1ccc(OC(=O)CCCC[C@@H]2SCC3NC(=O)NC23)cc1 |
| IUPAC Name | (4-nitrophenyl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate |
| Molecular Weight (g/mol) | 365.4 |
| Formula Weight | 365.1045 |