Learn More
Biotin X, TRC
Product Code. 30335400
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335400 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335400 Supplier Toronto Research Chemicals Supplier No. TRCB395670500MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Biotinylcaproic Acid
CAS 72040-64-3
Molecular Formula C16 H27 N3 O4 S
Quantity 500 mg
Synonym N-Biotinylcaproic Acid, 6-[[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]hexanoic Acid, 6-[(Biotinyl)amino]hexanoic acid, 1H-Thieno[3,4-d]imidazole, Hexanoic Acid Deriv., 6-(Biotinylamino)caproic Acid, 6-Biotinamidocaproic Acid
InChI Formula InChI=1S/C16H27N3O4S/c20-13(17-9-5-1-2-8-14(21)22)7-4-3-6-12-15-11(10-24-12)18-16(23)19-15/h11-12,15H,1-10H2,(H,17,20)(H,21,22)(H2,18,19,23)/t11-,12-,15-/m0/s1
SMILES OC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC Name 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoic acid
Molecular Weight (g/mol) 357.47
Formula Weight 357.1722