Learn More
Melford bis-MSB Scintillation Grade
Product Code. 30017920
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017920 25 g 25g
30017919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017920 Supplier Melford Supplier No. 111077
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High solubility secondary fluor.
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A
Specifications
Chemical Name or Material bis-MSB, Scintillation Grade
Quantity 25 g