Learn More
Bisphenol A-2,2',6,6'-d4, CDN
Product Code. 30321911
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321911 0.05 g 0.05g
30309786 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321911 Supplier CDN Supplier No. CDND29640.05G
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bisphenol A-2,2',6,6'-d4
CAS 102438-62-0
Molecular Formula C15 D4 H12 O2
Quantity 0.05 g
Synonym Phen-3,5-d2-ol, 4,4'-(1-methylethylidene)bis- (9 CI), Bisphenol A-2,2',6,6'-d4, Bisphenol A-2,2',6,6'-D4, 4,4'-(1-Methylethylidene)bisphen-3,5-d2-ol
InChI Formula InChI=1 S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17 H,1-2H3/i3D,4 D,5 D,6 D
SMILES [2 H]c1cc(O)cc([2 H])c1C(C)(C)c2c([2 H])cc(O)cc2[2 H]
IUPAC Name 3,5-dideuterio-4-[2-(2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol
Molecular Weight (g/mol) 232.311
Formula Weight 232.1401 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity