Learn More
STEMCELL Technologies BIT 9500 Serum Substitute
Product Code. 17189951
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17189951 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17189951 Supplier STEMCELL Technologies Supplier No. 09500
 Restricted Item
Expédition estimée: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Serum substitute for culturing human and mouse hematopoietic cells
  • BIT 9500 Serum Substitute has been developed for use in applications where a serum-free culture medium of defined composition is required
  • This product contains bovine serum albumin (BSA), insulin, and transferrin (BIT) in Iscove's MDM
  • It contains pre-screened batches of BSA that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations
  • It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions
Specifications
Cell Type Hematopoietic Stem and Progenitor Cells; Hybridomas; Other; Pluripotent Stem Cells
Quantity 100 mL
For Use With (Application) Cell Culture